siRNA Details
| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007016.1 | NP_001007017.1 | 7 | 922 | ||
| XM_008762902.1 | XP_008761124.1 | 8 | 2423 | |||
| GenBank mRNA | AJ629845.1 | - | 7 | 922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sspo| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007016.1 | NP_001007017.1 | 7 | 922 | ||
| XM_008762902.1 | XP_008761124.1 | 8 | 2423 | |||
| GenBank mRNA | AJ629845.1 | - | 7 | 922 | ||