siRNA Details
| Gene Symbol: Hmgn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007020.2 | NP_001007021.1 | 5 | 431 | ||
| GenBank mRNA | BC082036.1 | - | 5 | 386 | 7130894 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmgn3| Gene Symbol: Hmgn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007020.2 | NP_001007021.1 | 5 | 431 | ||
| GenBank mRNA | BC082036.1 | - | 5 | 386 | 7130894 | |