siRNA Details
| Gene Symbol: RESP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007089.3 | NP_001007090.3 | 6 | 615 | ||
| XM_011511165.2 | XP_011509467.1 | 4 | 478 | |||
| XM_011511167.2 | XP_011509469.1 | 5 | 929 | |||
| XM_017004079.1 | XP_016859568.1 | 5 | 772 | |||
| GenBank mRNA | AF437883.3 | - | 6 | 615 | ||
| DQ227570.1 | - | 4 | 686 | |||

RESP18
Genomic Map