siRNA Details
| Gene Symbol: FSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007122.3 | NP_001007123.1 | 7 | 1388 | ||
| NM_001281805.1 | NP_001268734.1 | 8 | 1505 | |||
| NM_001281806.1 | NP_001268735.1 | 7 | 1388 | |||
| XM_005272425.4 | XP_005272482.1 | 8 | 3131 | |||
| XM_011521235.2 | XP_011519537.1 | 7 | 2902 | |||
| XM_017021924.1 | XP_016877413.1 | 8 | 3131 | |||
| GenBank mRNA | AK122875.1 | - | 7 | 1387 | ||
| AL833295.1 | - | 7 | 1388 | |||
| BC130569.1 | - | 7 | 1372 | 40146600 | ||
| BC144175.1 | - | 8 | 1489 | 9052695 | ||
| BC144181.1 | - | 7 | 1372 | 9052701 | ||

FSD2
Genomic Map