siRNA Details
| Gene Symbol: Tob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007146.1 | NP_001007147.1 | 2 | 346 | ||
| GenBank mRNA | AY724519.1 | - | 2 | 549 | ||
| BC085117.1 | - | 2 | 385 | 7107186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tob2| Gene Symbol: Tob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007146.1 | NP_001007147.1 | 2 | 346 | ||
| GenBank mRNA | AY724519.1 | - | 2 | 549 | ||
| BC085117.1 | - | 2 | 385 | 7107186 | ||