siRNA Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007148.2 | NP_001007149.2 | 6 | 525 | ||
| XM_006231529.2 | XP_006231591.1 | 6 | 1108 | |||
| XM_006231530.2 | XP_006231592.1 | 5 | 519 | |||
| XM_006231531.2 | XP_006231593.1 | 5 | 485 | |||
| XM_006231535.2 | XP_006231597.1 | 5 | 370 | |||
| XM_006231536.2 | XP_006231598.1 | 5 | 452 | |||
| XM_006231537.2 | XP_006231599.1 | 5 | 999 | |||
| XM_006231538.2 | XP_006231600.1 | 4 | 409 | |||
| XM_006231539.2 | XP_006231601.1 | 4 | 1155 | |||
| XM_006231541.2 | XP_006231603.1 | 4 | 869 | |||
| XM_006231543.2 | XP_006231605.1 | 3 | 1077 | |||
| GenBank mRNA | BC085125.1 | - | 5 | 346 | 7131883 | |
| CB584165.1 | - | 6 | 538 | |||
| EV771066.1 | - | 6 | 465 | |||

Btrc
Genomic Map