siRNA Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007148.2 | NP_001007149.2 | 13 | 1653 | ||
| XM_006231529.2 | XP_006231591.1 | 13 | 2236 | |||
| XM_006231530.2 | XP_006231592.1 | 12 | 1647 | |||
| XM_006231531.2 | XP_006231593.1 | 12 | 1613 | |||
| XM_006231535.2 | XP_006231597.1 | 12 | 1498 | |||
| XM_006231536.2 | XP_006231598.1 | 12 | 1580 | |||
| XM_006231537.2 | XP_006231599.1 | 12 | 2127 | |||
| XM_006231538.2 | XP_006231600.1 | 11 | 1537 | |||
| XM_006231539.2 | XP_006231601.1 | 11 | 2283 | |||
| XM_006231541.2 | XP_006231603.1 | 11 | 1997 | |||
| XM_006231543.2 | XP_006231605.1 | 10 | 2205 | |||
| GenBank mRNA | BC085125.1 | - | 12 | 1474 | 7131883 | |

Btrc
Genomic Map