siRNA Details
| Gene Symbol: Itpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007235.2 | NP_001007236.2 | 19 | 2154 | ||
| NM_001270596.1 | NP_001257525.1 | 18 | 2109 | |||
| NM_001270597.1 | NP_001257526.1 | 19 | 2154 | |||
| XM_006236987.2 | XP_006237049.1 | 19 | 2192 | |||
| XM_008763167.1 | XP_008761389.1 | 19 | 2590 | |||
| XM_008763168.1 | XP_008761390.1 | 19 | 2590 | |||
| XM_008763169.1 | XP_008761391.1 | 19 | 2590 | |||
| XM_008763170.1 | XP_008761392.1 | 19 | 2590 | |||
| XM_008763171.1 | XP_008761393.1 | 19 | 2589 | |||
| XM_008763172.1 | XP_008761394.1 | 19 | 2589 | |||
| XM_008763173.1 | XP_008761395.1 | 18 | 2544 | |||
| XM_008763174.1 | XP_008761396.1 | 19 | 2589 | |||
| XM_008763175.1 | XP_008761397.1 | 18 | 2544 | |||
| XM_008763176.1 | XP_008761398.1 | 19 | 2588 | |||
| XM_008763177.1 | XP_008761399.1 | 18 | 2544 | |||
| XM_008763178.1 | XP_008761400.1 | 18 | 2543 | |||
| GenBank mRNA | GQ233032.1 | - | 17 | 1916 | ||
| J05510.1 | - | 19 | 2245 | |||

Itpr1
Genomic Map