siRNA Details
Gene Symbol: LOC107986113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007620.1 | XP_016863109.1 | 2 | 423 |
Gene Symbol: ERVK-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN345079.1 | - | 1 | 560 |
Gene Symbol: LOC105374013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513341.2 | XP_011511643.1 | 2 | 911 |
Gene Symbol: LOC107986062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007615.1 | XP_016863104.1 | 2 | 432 |
