siRNA Details
| Gene Symbol: ERVK-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FN806847.1 | - | 1 | 31 | ||
| Gene Symbol: ERVK-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FN806835.1 | - | 1 | 31 | ||
| Gene Symbol: ERVK-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FN806842.1 | - | 1 | 31 | ||
| Gene Symbol: ERVK-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FN806829.1 | - | 1 | 31 | ||
| Gene Symbol: LOC107986062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007615.1 | XP_016863104.1 | 2 | 1710 | ||
| Gene Symbol: LOC107986113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007620.1 | XP_016863109.1 | 2 | 1704 | ||

ERVK-23
Genomic Map