siRNA Details
| Gene Symbol: Os9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007265.1 | NP_001007266.1 | 15 | 2296 | ||
| XM_006241461.1 | XP_006241523.1 | 14 | 2152 | |||
| GenBank mRNA | BC085907.1 | - | 15 | 2296 | 7096059 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Os9| Gene Symbol: Os9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007265.1 | NP_001007266.1 | 15 | 2296 | ||
| XM_006241461.1 | XP_006241523.1 | 14 | 2152 | |||
| GenBank mRNA | BC085907.1 | - | 15 | 2296 | 7096059 | |