siRNA Details
| Gene Symbol: NWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007525.3 | NP_001007526.3 | 19 | 6476 | ||
| NM_001290355.1 | NP_001277284.1 | 21 | 7234 | |||
| XM_011527929.1 | XP_011526231.1 | 21 | 6766 | |||
| XM_011527930.2 | XP_011526232.1 | 21 | 7252 | |||
| XM_011527931.2 | XP_011526233.1 | 20 | 7032 | |||
| XM_011527934.1 | XP_011526236.1 | 16 | 4971 | |||
| XM_017026669.1 | XP_016882158.1 | 22 | 7319 | |||
| GenBank mRNA | AK026798.1 | - | 1 | 992 | ||
| AK090685.1 | - | 3 | 2200 | |||
| BX648940.1 | - | 22 | 7234 | |||
| CR749794.1 | - | 19 | 6472 | |||

NWD1
Genomic Map