siRNA Details
| Gene Symbol: NWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007525.3 | NP_001007526.3 | 19 | 5787 | ||
| NM_001290355.1 | NP_001277284.1 | 21 | 6545 | |||
| XM_011527929.1 | XP_011526231.1 | 21 | 6077 | |||
| XM_011527930.2 | XP_011526232.1 | 21 | 6563 | |||
| XM_011527931.2 | XP_011526233.1 | 20 | 6343 | |||
| XM_011527934.1 | XP_011526236.1 | 16 | 4282 | |||
| XM_017026669.1 | XP_016882158.1 | 22 | 6630 | |||
| GenBank mRNA | AK026798.1 | - | 1 | 303 | ||
| AK090685.1 | - | 3 | 1511 | |||
| BC117698.1 | - | 19 | 5541 | 40073616 | ||
| BX648940.1 | - | 22 | 6545 | |||
| CR749794.1 | - | 19 | 5783 | |||

NWD1
Genomic Map