siRNA Details
| Gene Symbol: USP6NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080491.3 | NP_001073960.1 | 3 | 189 | ||
| NM_014688.3 | NP_055503.1 | 4 | 520 | |||
| XM_006717542.3 | XP_006717605.1 | 5 | 346 | |||
| XM_011519762.2 | XP_011518064.1 | 6 | 679 | |||
| XM_017016971.1 | XP_016872460.1 | 5 | 353 | |||
| XM_017016972.1 | XP_016872461.1 | 4 | 312 | |||
| XM_017016973.1 | XP_016872462.1 | 3 | 3516 | |||
| GenBank mRNA | AB449916.1 | - | 2 | 90 | ||
| AK292944.1 | - | 4 | 310 | |||
| BC042943.2 | - | 4 | 358 | 5296060 | ||
| CF121539.1 | - | 3 | 196 | |||
| D13644.1 | - | 4 | 369 | |||
| DB457665.1 | - | 4 | 411 | |||

USP6NL
Genomic Map