siRNA Details
| Gene Symbol: NKAPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007531.2 | NP_001007532.1 | 1 | 622 | ||
| GenBank mRNA | AF085870.1 | - | 1 | 375 | ||
| BC038240.1 | - | 1 | 552 | 5766894 | ||
| BC101681.1 | - | 1 | 580 | 8069187 | ||
| BC101685.1 | - | 1 | 580 | 8069191 | ||

NKAPL
Genomic Map