siRNA Details
| Gene Symbol: LINC00167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024233.1 | 1 | 1408 | |||
| GenBank mRNA | AK055956.1 | - | 1 | 1408 | ||
| BC137306.1 | - | 1 | 1094 | |||
| BC137307.1 | - | 1 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00167| Gene Symbol: LINC00167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024233.1 | 1 | 1408 | |||
| GenBank mRNA | AK055956.1 | - | 1 | 1408 | ||
| BC137306.1 | - | 1 | 1094 | |||
| BC137307.1 | - | 1 | 1094 | |||