siRNA Details
| Gene Symbol: Fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007554.1 | NP_001007555.1 | 9 | 1315 | ||
| XM_008764273.1 | XP_008762495.1 | 9 | 3539 | |||
| XM_008764274.1 | XP_008762496.1 | 8 | 2702 | |||
| XM_008764275.1 | XP_008762497.1 | 9 | 2771 | |||
| XM_008764276.1 | XP_008762498.1 | 9 | 2922 | |||
| GenBank mRNA | BC086398.1 | - | 9 | 1315 | 7303205 | |

Fblim1
Genomic Map