siRNA Details
| Gene Symbol: Emr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007557.1 | NP_001007558.1 | 7 | 784 | ||
| XM_006244280.2 | XP_006244342.1 | 7 | 805 | |||
| XM_008766759.1 | XP_008764981.1 | 6 | 840 | |||
| GenBank mRNA | AY686632.1 | - | 7 | 784 | ||
More Information
Set Membership: |
|

Emr1
Genomic Map