siRNA Details
| Gene Symbol: Zfp251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007568.2 | NP_001007569.1 | 4 | 2844 | ||
| GenBank mRNA | AK047299.1 | - | 120 | |||
| AK140082.1 | - | 4 | 2835 | |||
| AK153967.1 | - | 3 | 2684 | |||
| BC041112.1 | - | 1280 | ||||
| BC059071.1 | - | 4 | 2844 | 6826152 | ||

Zfp251
Genomic Map