siRNA Details
| Gene Symbol: Pnmal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007569.1 | NP_001007570.1 | 2 | 424 | ||
| GenBank mRNA | AK079661.1 | - | 2 | 540 | ||
| BC048228.1 | - | 2 | 424 | 6310552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnmal1| Gene Symbol: Pnmal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007569.1 | NP_001007570.1 | 2 | 424 | ||
| GenBank mRNA | AK079661.1 | - | 2 | 540 | ||
| BC048228.1 | - | 2 | 424 | 6310552 | ||