siRNA Details
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007578.2 | NP_001007579.1 | 4 | 829 | ||
GenBank mRNA | AK148507.1 | - | 4 | 829 | ||
BC027264.1 | - | 4 | 822 | 3492516 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007578.2 | NP_001007579.1 | 4 | 829 | ||
GenBank mRNA | AK148507.1 | - | 4 | 829 | ||
BC027264.1 | - | 4 | 822 | 3492516 |