siRNA Details
Gene Symbol: Fndc3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007580.1 | NP_001007581.1 | 26 | 4014 | ||
GenBank mRNA | BC076625.1 | - | 26 | 4014 | 30641582 | |
BC085176.1 | - | 26 | 4012 | 30626050 | ||
DQ192038.1 | - | 26 | 4059 |
