siRNA Details
| Gene Symbol: Gm11541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007584.2 | NP_001007585.1 | 2 | 1578 | ||
| GenBank mRNA | AK082500.1 | - | 2 | 1579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11541| Gene Symbol: Gm11541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007584.2 | NP_001007585.1 | 2 | 1578 | ||
| GenBank mRNA | AK082500.1 | - | 2 | 1579 | ||