siRNA Details
| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 9 | 4490 | ||
| GenBank mRNA | AK091416.1 | - | 6 | 2261 | ||
| Gene Symbol: KIAA0408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014702.4 | NP_055517.3 | 3 | 796 | ||
| GenBank mRNA | AB007868.1 | - | 1 | 1189 | ||
| AK091424.1 | - | 4 | 1152 | |||
| AK125983.1 | - | 4 | 1305 | |||
| BC018396.1 | - | 3 | 634 | 4184033 | ||

SOGA3
Genomic Map