siRNA Details
| Gene Symbol: BB014433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077224.1 | - | 2 | 1559 | ||
| BC139040.1 | - | 1 | 1439 | 8862062 | ||
| BC145521.1 | - | 2 | 1187 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB014433| Gene Symbol: BB014433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077224.1 | - | 2 | 1559 | ||
| BC139040.1 | - | 1 | 1439 | 8862062 | ||
| BC145521.1 | - | 2 | 1187 | |||