siRNA Details
| Gene Symbol: CEP104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014704.3 | NP_055519.1 | 4, 5 | 793 | ||
| XM_005244815.4 | XP_005244872.1 | 5, 6 | 893 | |||
| XM_011542473.2 | XP_011540775.1 | 5, 6 | 1093 | |||
| XM_011542474.2 | XP_011540776.1 | 4, 5 | 811 | |||
| XM_011542475.2 | XP_011540777.1 | 5, 6 | 1093 | |||
| XM_011542476.2 | XP_011540778.1 | 5, 6 | 1093 | |||
| XM_011542477.2 | XP_011540779.1 | 5, 6 | 1093 | |||
| XM_011542478.1 | XP_011540780.1 | 5, 6 | 1093 | |||
| XM_017002917.1 | XP_016858406.1 | 5, 6 | 1075 | |||
| XM_017002918.1 | XP_016858407.1 | 4, 5 | 793 | |||
| XM_017002919.1 | XP_016858408.1 | 4, 5 | 771 | |||
| GenBank mRNA | AB011134.1 | - | 4, 5 | 747 | ||
| AK308191.1 | - | 4, 5 | 752 | |||
| BC001640.2 | - | 4, 5 | 758 | |||
| BC047450.1 | - | 4, 5 | 758 | 5297035 | ||
| BC050721.1 | - | 4, 5 | 774 | |||
| DA081362.1 | - | 2, 3 | 235 | |||

CEP104
Genomic Map