siRNA Details
| Gene Symbol: Pcbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007601.2 | NP_001007602.1 | 4 | 327 | ||
| XM_006256414.1 | XP_006256476.1 | 5 | 585 | |||
| GenBank mRNA | DV724713.1 | - | 2 | 136 | ||
| FM060466.1 | - | 4 | 327 | |||
| FM067129.1 | - | 4 | 304 | |||
| M83740.1 | - | 4 | 523 | |||

Pcbd1
Genomic Map