siRNA Details
| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007602.1 | NP_001007603.1 | 5, 6 | 654 | ||
| GenBank mRNA | AB008807.1 | - | 5, 6 | 636 | ||
| BC079363.1 | - | 5, 6 | 654 | 7109248 | ||
| FJ179405.1 | - | 5, 6 | 572 | |||
| FQ218105.1 | - | 5, 6 | 629 | |||
| FQ220742.1 | - | 5, 6 | 596 | |||

Gsto1
Genomic Map