siRNA Details
| Gene Symbol: Mfap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007609.1 | NP_001007610.1 | 2 | 290 | ||
| XM_006246379.2 | XP_006246441.1 | 2 | 451 | |||
| GenBank mRNA | BC079064.1 | - | 2 | 290 | 7121173 | |
| EU000470.1 | - | 1 | 272 | |||

Mfap3
Genomic Map