siRNA Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007616.1 | NP_001007617.1 | 2 | 134 | ||
| XM_008769220.1 | XP_008767442.1 | 1 | 71 | |||
| GenBank mRNA | BC081901.1 | - | 2 | 134 | 7110808 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wsb2| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007616.1 | NP_001007617.1 | 2 | 134 | ||
| XM_008769220.1 | XP_008767442.1 | 1 | 71 | |||
| GenBank mRNA | BC081901.1 | - | 2 | 134 | 7110808 | |