siRNA Details
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007618.1 | NP_001007619.1 | 2 | 494 | ||
GenBank mRNA | BC083739.1 | - | 2 | 494 | 7191744 | |
FQ216834.1 | - | 2 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007618.1 | NP_001007619.1 | 2 | 494 | ||
GenBank mRNA | BC083739.1 | - | 2 | 494 | 7191744 | |
FQ216834.1 | - | 2 | 261 |