siRNA Details
| Gene Symbol: Pdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007620.1 | NP_001007621.1 | 6 | 690 | ||
| XM_006251754.1 | XP_006251816.1 | 7 | 550 | |||
| GenBank mRNA | BC079137.1 | - | 6 | 690 | 7127701 | |
| FQ212778.1 | - | 6 | 379 | |||
| FQ214416.1 | - | 6 | 379 | |||
| FQ234205.1 | - | 6 | 379 | |||

Pdhb
Genomic Map