siRNA Details
| Gene Symbol: Mettl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007623.1 | NP_001007624.1 | 4 | 672 | ||
| XM_006252582.2 | XP_006252644.1 | 3 | 567 | |||
| XM_006252583.2 | XP_006252645.1 | 3 | 570 | |||
| XM_008770988.1 | XP_008769210.1 | 3 | 567 | |||
| GenBank mRNA | BC079309.1 | - | 4 | 672 | 7134747 | |
| FQ211422.1 | - | 4 | 680 | |||

Mettl6
Genomic Map