siRNA Details
| Gene Symbol: Epdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007625.1 | NP_001007626.1 | 2, 3 | 695 | ||
| GenBank mRNA | BC083701.1 | - | 2, 3 | 695 | 7190104 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epdr1| Gene Symbol: Epdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007625.1 | NP_001007626.1 | 2, 3 | 695 | ||
| GenBank mRNA | BC083701.1 | - | 2, 3 | 695 | 7190104 | |