siRNA Details
| Gene Symbol: Stard6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007627.1 | NP_001007628.1 | 6 | 602 | ||
| XM_006254921.2 | XP_006254983.1 | 7 | 824 | |||
| XM_008772111.1 | XP_008770333.1 | 7 | 672 | |||
| XM_008772112.1 | XP_008770334.1 | 6 | 627 | |||
| GenBank mRNA | AY555189.1 | - | 7 | 772 | ||
| BC078976.1 | - | 6 | 602 | 7111945 | ||

Stard6
Genomic Map