siRNA Details
| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007628.2 | NP_001007629.1 | 2 | 1837 | ||
| GenBank mRNA | BC083622.1 | - | 3 | 1602 | 7134100 | |
| DV723039.1 | - | 1 | 720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxxc5| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007628.2 | NP_001007629.1 | 2 | 1837 | ||
| GenBank mRNA | BC083622.1 | - | 3 | 1602 | 7134100 | |
| DV723039.1 | - | 1 | 720 | |||