siRNA Details
| Gene Symbol: mrpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007637.1 | NP_001007638.1 | 6 | 1049 | ||
| GenBank mRNA | BC082018.1 | - | 6 | 1049 | 7132470 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpl24| Gene Symbol: mrpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007637.1 | NP_001007638.1 | 6 | 1049 | ||
| GenBank mRNA | BC082018.1 | - | 6 | 1049 | 7132470 | |