siRNA Details
| Gene Symbol: TCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206938.1 | NP_001193867.1 | 2 | 283 | ||
| NM_014719.2 | NP_055534.1 | 2 | 243 | |||
| XM_005250074.3 | XP_005250131.1 | 2 | 283 | |||
| XM_005250075.2 | XP_005250132.1 | 2 | 285 | |||
| XM_005250076.3 | XP_005250133.1 | 2 | 320 | |||
| XM_005250077.2 | XP_005250134.1 | 3 | 449 | |||
| XM_006716191.2 | XP_006716254.1 | 2 | 249 | |||
| XM_006716192.3 | XP_006716255.1 | 2 | 392 | |||
| XM_017012833.1 | XP_016868322.1 | 2 | 315 | |||
| XM_017012834.1 | XP_016868323.1 | 2 | 285 | |||
| XM_017012835.1 | XP_016868324.1 | 3 | 449 | |||
| XM_017012836.1 | XP_016868325.1 | 2 | 249 | |||
| GenBank mRNA | AB018281.1 | - | 2 | 243 | ||
| AK001640.1 | - | 2 | 319 | |||
| AK291605.1 | - | 2 | 317 | |||
| BC000609.2 | - | 2 | 224 | 3346754 | ||
| DA196926.1 | - | 2 | 288 | |||
| DA317873.1 | - | 2 | 283 | |||

TCAF1
Genomic Map