siRNA Details
Gene Symbol: Phospho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007642.1 | NP_001007643.1 | 4 | 638 | ||
GenBank mRNA | BC081926.1 | - | 4 | 638 | 7113082 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Phospho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007642.1 | NP_001007643.1 | 4 | 638 | ||
GenBank mRNA | BC081926.1 | - | 4 | 638 | 7113082 |