siRNA Details
| Gene Symbol: Mturn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007645.1 | NP_001007646.1 | 3 | 903 | ||
| XM_008762880.1 | XP_008761102.1 | 4 | 550 | |||
| GenBank mRNA | BC083875.1 | - | 3 | 903 | 7129817 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mturn| Gene Symbol: Mturn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007645.1 | NP_001007646.1 | 3 | 903 | ||
| XM_008762880.1 | XP_008761102.1 | 4 | 550 | |||
| GenBank mRNA | BC083875.1 | - | 3 | 903 | 7129817 | |