siRNA Details
| Gene Symbol: Cdca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007648.1 | NP_001007649.1 | 6 | 1321 | ||
| XM_006237341.2 | XP_006237403.1 | 6 | 905 | |||
| XM_006237342.2 | XP_006237404.1 | 5 | 1338 | |||
| XM_008763281.1 | XP_008761503.1 | 6 | 1043 | |||
| GenBank mRNA | BC079204.1 | - | 6 | 1321 | 7131776 | |
| FQ209817.1 | - | 3 | 466 | |||
| FQ235136.1 | - | 6 | 859 | |||

Cdca3
Genomic Map