siRNA Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007652.2 | NP_001007653.1 | 4 | 1514 | ||
| GenBank mRNA | BC081988.1 | - | 4 | 1514 | 7129121 | |
| BC083800.1 | - | 4 | 1514 | 7108773 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pole3| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007652.2 | NP_001007653.1 | 4 | 1514 | ||
| GenBank mRNA | BC081988.1 | - | 4 | 1514 | 7129121 | |
| BC083800.1 | - | 4 | 1514 | 7108773 | ||