siRNA Details
| Gene Symbol: Agtrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007654.1 | NP_001007655.1 | 5 | 1951 | ||
| GenBank mRNA | BC082019.1 | - | 5 | 1951 | 7129329 | |
| FQ231399.1 | - | 1 | 811 | |||
| FQ231638.1 | - | 1 | 1188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agtrap| Gene Symbol: Agtrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007654.1 | NP_001007655.1 | 5 | 1951 | ||
| GenBank mRNA | BC082019.1 | - | 5 | 1951 | 7129329 | |
| FQ231399.1 | - | 1 | 811 | |||
| FQ231638.1 | - | 1 | 1188 | |||