siRNA Details
| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007658.1 | NP_001007659.1 | 2 | 1206 | ||
| GenBank mRNA | BC083742.1 | - | 2 | 1206 | 7191975 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Them6| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007658.1 | NP_001007659.1 | 2 | 1206 | ||
| GenBank mRNA | BC083742.1 | - | 2 | 1206 | 7191975 | |