siRNA Details
Gene Symbol: Atg101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007659.2 | NP_001007660.2 | 4 | 938 | ||
NM_001271114.1 | NP_001258043.1 | 3 | 763 | |||
XM_006242324.2 | XP_006242386.1 | 4 | 901 | |||
GenBank mRNA | BC079170.1 | - | 4 | 916 | 7129890 |
