siRNA Details
| Gene Symbol: Atg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007659.2 | NP_001007660.2 | 4 | 1174 | ||
| NM_001271114.1 | NP_001258043.1 | 3 | 999 | |||
| XM_006242324.2 | XP_006242386.1 | 4 | 1137 | |||
| GenBank mRNA | BC079170.1 | - | 4 | 1152 | 7129890 | |

Atg101
Genomic Map