siRNA Details
| Gene Symbol: Arcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007662.1 | NP_001007663.1 | 10 | 1590 | ||
| GenBank mRNA | BC081979.1 | - | 10 | 1590 | 7128203 | |
| FQ229375.1 | - | 4 | 455 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arcn1| Gene Symbol: Arcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007662.1 | NP_001007663.1 | 10 | 1590 | ||
| GenBank mRNA | BC081979.1 | - | 10 | 1590 | 7128203 | |
| FQ229375.1 | - | 4 | 455 | |||