siRNA Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007665.1 | NP_001007666.1 | 5 | 779 | ||
| XM_006243726.2 | XP_006243788.1 | 5 | 745 | |||
| XM_006243727.2 | XP_006243789.1 | 5 | 1046 | |||
| XM_008766501.1 | XP_008764723.1 | 5 | 735 | |||
| GenBank mRNA | BC083732.1 | - | 5 | 779 | 7191345 | |

Tex264
Genomic Map