siRNA Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282533.1 | NP_001269462.1 | 3 | 532 | ||
| XM_005260890.3 | XP_005260947.1 | 3 | 476 | |||
| XM_005260892.1 | XP_005260949.1 | 3 | 532 | |||
| XM_005260893.2 | XP_005260950.1 | 3 | 445 | |||
| XM_011529407.2 | XP_011527709.1 | 3 | 476 | |||
| XM_011529408.2 | XP_011527710.1 | 3 | 1514 | |||
| XM_011529409.2 | XP_011527711.1 | 3 | 1934 | |||
| GenBank mRNA | AB011124.1 | - | 1 | 1560 | ||
| BC038860.1 | - | 3 | 488 | 5260571 | ||

LZTS3
Genomic Map