siRNA Details
| Gene Symbol: Sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007667.2 | NP_001007668.1 | 6 | 916 | ||
| XM_008773228.1 | XP_008771450.1 | 6 | 1916 | |||
| XM_008773229.1 | XP_008771451.1 | 6 | 1312 | |||
| XM_008773230.1 | XP_008771452.1 | 7 | 951 | |||
| GenBank mRNA | BC060588.1 | - | 6 | 918 | 6921572 | |
| FQ223333.1 | - | 1 | 192 | |||
| FQ229984.1 | - | 6 | 800 | |||

Sat1
Genomic Map